Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IKK beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4535SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4535SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4535SP Supplier R&D Systems Supplier No. AF4535SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

IKK beta Polyclonal specifically detects IKK beta in Human samples. It is validated for Western Blot.

Specifications

Antigen IKK beta
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O14920
Gene Alias EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKB, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta
Gene Symbols IKBKB
Host Species Sheep
Immunogen E. coli-derived recombinant human IKK beta Gly374-Ser756 Accession # O14920
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3551
Test Specificity Detects endogenous human IKK beta in Western blots. In Western blots, this antibody does not cross-react with recombinant human IKK alpha , IKK gamma , or IKK epsilon .
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.