Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-1 alpha/IL-1F1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB200R100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB200R100 100 μg
MAB200RSP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB200R100 Supplier R&D Systems Supplier No. MAB200R100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-1 alpha/IL-1F1 Monoclonal specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-1 alpha/IL-1F1
Applications Neutralization, Immunocytochemistry, ELISA
Classification Monoclonal
Clone 4414R
Conjugate Unconjugated
Dilution Neutralization 0.05-0.15 ug/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01583
Gene Alias BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha
Gene Symbols IL1A
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-1 alpha/IL-1F1 Ser113-Ala271 Accession # P01583
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3552
Test Specificity Detects human IL-1 alpha/IL-1F1 in ELISAs and western blots. In ELISAs, this antibody does not cross-react with recombinant human (rh) IL-1 beta , -2, -3, -4, -5, -6, -7, -8, -9, -10, -11, -12, -13, rmIL-1 alpha , -1 beta , -3, -4, -5, -6, -7, -9, or -13.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.