Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-1 alpha/IL-1F1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $409.00
Specifications
Antigen | IL-1 alpha/IL-1F1 |
---|---|
Clone | 1042710 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2002SP
![]() |
R&D Systems
MAB2002SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2002100
![]() |
R&D Systems
MAB2002100 |
100 μg |
Each for $409.00
|
|
|||||
Description
IL-1 alpha/IL-1F1 Monoclonal antibody specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for ImmunohistochemistrySpecifications
IL-1 alpha/IL-1F1 | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
E. coli-derived recombinant human IL-1 alpha/IL-1F1, Ser113-Ala271, Accession # Q53QF9 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1042710 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q53QF9 | |
3552 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title