Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-1 alpha/IL-1F1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: R&D Systems BAF200
This item is not returnable.
View return policy
Description
IL-1 alpha/IL-1F1 Polyclonal specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Western Blot, Immunocytochemistry, ELISA Detection (Matched Antibody Pair).Specifications
IL-1 alpha/IL-1F1 | |
Polyclonal | |
Biotin | |
Q53QF9 | |
IL1A | |
E. coli-derived recombinant human IL-1 alpha/IL-1F1 (R&D Systems, Catalog # 200-LA) Ser113-Ala271 Accession # Q53QF9 | |
50 μg | |
Primary | |
Detects human IL-1 alpha/IL-1F1 in ELISAs and Western blots. In sandwich immunoassays, less than 0.05% cross-reactivity with recombinant mouse IL-1 alpha , recombinant porcine IL-1 alpha , recombinant rat IL-1 alpha , recombinant human (rh) IL-1 sRI, and rhIL-1 sRII is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
Goat | |
Affinity Purified | |
RUO | |
3552 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction