Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human IL-1 alpha/IL-1F1 ELISA Kit (Colorimetric)

Click to view available options
Quantity:
96 Tests
Description
The Human IL-1 alpha/IL-1F1 ELISA Kit (Colorimetric) measures human IL-1 alpha/IL-1F1 in cell culture supernatant, serum, plasma (edta, citrate, heparin).
Specifications
Specifications
Assay Range | 1.6 - 100pg/mL |
Assay Sensitivity | 1.1pg/mL |
Product Type | ELISA Kit |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
Target Species | Human |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 3552 |
Gene Symbol | IL1A |
Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to human IL-1alpha, Biotin-Conjugate anti-human IL-1alpha polyclonal antibody, Streptavidin-HRP, Human IL-1alpha Standard lyophilized, 200 pg/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
Quantity | 96 Tests |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction