Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-1 beta/IL-1F2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB601500
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Western Blot, Simple Western, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).Specifications
IL-1 beta/IL-1F2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IL1B | |
E. coli-derived recombinant human IL-1 beta/IL-1F2 | |
500 μg | |
Primary | |
Detects human IL-1 beta/IL-1F2 in sandwich ELISAs and Western blots. In sandwich ELISAs, less than 4% cross-reactivity with recombinant rat (rr) IL-1 beta and less than 0.1% with recombinant porcine (rp) IL-1 beta , recombinant human IL-1 alpha , rpIL-1 alpha , rrIL-1 alpha , recombinant mouse (rm) IL-1 alpha , and rmIL-1 beta is observed. | |
Human | |
Supernatant |
Western Blot, Neutralization, Immunocytochemistry, ELISA | |
2805 | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Neutralization 0.05-0.2 ug/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3553 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction