Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-1 beta/IL-1F2 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC201A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC201A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC201A Supplier R&D Systems Supplier No. IC201A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-1 beta/IL-1F2
Applications Flow Cytometry
Classification Monoclonal
Clone 8516
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P01584
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Gene Symbols IL1B
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-1 beta/IL-1F2 aa 117-269 Accession # P01584
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3553
Test Specificity Detects human IL-1 beta/IL-1F2 in Western blots. Shows less than 5% cross-reactivity with recombinant mouse (rm) IL-1 beta and rpIL-1 beta and no cross-reactivity with rrIL-1 beta , rmIL-1 alpha , rhIL-1ra, rmIL-1ra, or rrIL-1 alpha .
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.