Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-1 beta/IL-1F2 Propeptide Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems MAB6964SP
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
IL-1 beta/IL-1F2 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P01584 | |
IL1B | |
E. coli-derived recombinant human IL-1 beta/IL-1F2 Propeptide Met1-Asp116 Accession # P01584 | |
25 μg | |
Primary | |
Detects human IL-1 beta/IL-1F2 Propeptide in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 alpha, rhIL-1F7, rhIL-1F10, rhIL-18, rhIL-36 alpha, rhIL-36 beta, IL-36 gamma, rhIL-36Ra, recombinant mouse IL-1Ra, or the mature peptides of recombinant IL-1 beta from canine, cotton rat, equine, feline, guinea pig, human, mouse, porcine, rabbit, rat, or rhesus is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
615417 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3553 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction