Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-1 beta/IL-1F2 QuantiGlo ELISA Kit

Catalog No. qlb00b
Click to view available options
Sufficient For:
1 x 96-well Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Human IL-1 beta/IL-1F2 QuantiGlo ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-1 beta/IL-1F2 QuantiGlo ELISA Kit quantifies human IL-1 beta/IL-1F2 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).

Specifications

Assay Range 1.40 - 1000pg/mL
Assay Sensitivity 0.55pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human IL-1 beta. This assay also recognizes natural and Recombinant rhesus macaque IL-1 beta. This assay cross-reacts 15.4% with Recombinant Human Pro-IL-1 beta , 1.0% with Recombinant Rat IL-1 beta , 0.6% with Recombinant Cotton Rat IL-1 beta .
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 3553
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym catabolin, IL-1, IL1 beta, IL-1 beta, IL-1 b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.