Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-1 beta/IL-1F2 Quantikine HS ELISA Kit
GSA_VASDP

Catalog No. SSLB00D Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SSLB00D 1 Pk. (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SSLB00D Supplier R&D Systems™ Supplier No. SSLB00D

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human IL-1 beta/IL-1F2 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-1 beta/IL-1F2 Quantikine HS ELISA Kit quantifies human IL-1 beta/IL-1F2 in serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), saliva (10 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type ELISA Kit
Quantity 1 Pk. (6 Plates)
Species Human
Conjugate Biotin
Binding Target IL-1 beta/IL-1F2
Assay Sensitivity 0.063 pg/mL
Assay Range 0.13 to 8 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 3553
Product Line R&D Systems
Sample Type Serum (100uL), EDTA Plasma (100uL), Heparin Plasma (100uL), Saliva (10uL)
Synonym catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Cytokines
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.