Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-1 beta/IL-1F2 (Research Grade Canakinumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | IL-1 beta/IL-1F2 |
---|---|
Clone | Hu110 |
Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10349SP
![]() |
R&D Systems
MAB10349SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10349100
![]() |
R&D Systems
MAB10349100 |
100 μg |
Each for $502.50
|
|
|||||
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
IL-1 beta/IL-1F2 | |
Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
3553 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Hu110 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Human | |
Human | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Human IL-1 beta/IL-1F2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects Human IL-1 beta/IL-1F2 in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Canakinumab. This product is for research use only. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title