Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-1 RAcP/IL-1 R3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB676R100
This item is not returnable.
View return policy
Description
IL-1 RAcP/IL-1 R3 Monoclonal specifically detects IL-1 RAcP/IL-1 R3 in Human samples. It is validated for Flow Cytometry.Specifications
IL-1 RAcP/IL-1 R3 | |
Monoclonal | |
Unconjugated | |
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3, FLJ37788, IL-1 R3, IL-1 receptor accessory protein, IL-1R3, IL-1R-3, IL1RAcP, IL-1RAcP, IL1RAP, interleukin 1 receptor accessory protein, interleukin-1 receptor accessory protein, interleukin-1 receptor accessory protein beta | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3556 | |
Human | |
Purified |
Flow Cytometry | |
89412R | |
Flow Cytometry 0.25 ug/10^6 cells | |
Q9NPH3 | |
IL1RAP | |
S. frugiperda insect ovarian cell line Sf21-derived recombinant human IL-1RAcP/IL-1 R3 Ser21-Glu359 Accession # Q9NPH3 | |
100 μg | |
Primary | |
Detects human IL-1RAcP/IL-1R3 in direct ELISAs. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction