Learn More
Human IL-1 RI Alexa Fluor 700-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | IL-1 RI |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Alias | CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 |
| Gene Symbols | IL1R1 |
| Host Species | Goat |
| Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-1 RI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.