Learn More
Human IL-1 RI PE-conjugated Antibody, R&D Systems™



Goat Polyclonal Antibody
$259.67 - $520.12
Specifications
| Antigen | IL-1 RI |
|---|---|
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | R-PE |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB269P025
![]() ![]() |
R&D Systems
FAB269P025 |
25 Tests |
Each for $259.67
|
|
|||||
FAB269P100
![]() |
R&D Systems
FAB269P100 |
100 Tests |
Each for $520.12
|
|
|||||
Description
IL-1 RI Polyclonal specifically detects IL-1 RI in Human samples. It is validated for Flow Cytometry.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
| IL-1 RI | |
| Flow Cytometry | |
| R-PE | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 3554 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-1 RI | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry 10 uL/10^6 cells | |
| Polyclonal | |
| Goat | |
| Human | |
| CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
| IL1R1 | |
| IgG | |
| Affinity Purified | |
| Detects human IL-1 RI in direct ELISAs and Western blots. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.