Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $538.00
Specifications
Antigen | IL-10 |
---|---|
Clone | 21204R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB91841SP
![]() |
R&D Systems
MAB91841SP |
25 μg |
Each for $241.00
|
|
|||||
MAB91841100
![]() |
R&D Systems
MAB91841100 |
100 μg |
Each for $538.00
|
|
|||||
Description
IL-10 Monoclonal specifically detects IL-10 in Human samples. It is validated for ELISA.Specifications
IL-10 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
P22301 | |
3586 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
21204R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
E. coli-derived recombinant human IL-10 Ser19-Asn178 Accession # P22301 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human IL-10 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-10 or recombinant rat IL-10 is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title