Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-10 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $526.00
Specifications
Antigen | IL-10 |
---|---|
Clone | 997935 |
Dilution | Immunohistochemistry 0.5-25 μg;/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB92101SP
![]() |
R&D Systems
MAB92101SP |
25 μg |
Each for $200.50
|
|
|||||
MB92101100
![]() |
R&D Systems
MAB92101100 |
100 μg |
Each for $526.00
|
|
|||||
Description
IL-10 Monoclonal specifically detects IL-10 in Human samples. It is validated for Immunohistochemistry.Specifications
IL-10 | |
Immunohistochemistry 0.5-25 μg;/mL | |
Monoclonal | |
Supernatant | |
Human | |
P22301 | |
3586.0 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
997935 | |
Immunohistochemistry | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
Human embryonic kidney cell, HEK293-derived human IL-10, Met1-Asn178, Accession No. P22301 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-10 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title