Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 12 publications
$227.50 - $2070.00
Specifications
Antigen | IL-10 |
---|---|
Clone | 25209 |
Dilution | Western Blot 5 ug/mL, Neutralization 0.005-0.03 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2171SP
![]() |
R&D Systems
MAB2171SP |
25 μg |
Each for $227.50
|
|
|||||
MAB2171100
![]() |
R&D Systems
MAB2171100 |
100 μg |
Each for $561.50
|
|
|||||
MAB2171500
![]() |
R&D Systems
MAB2171500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
IL-10 Monoclonal specifically detects IL-10 in Human samples. It is validated for Western Blot, Neutralization.Specifications
IL-10 | |
Western Blot 5 ug/mL, Neutralization 0.005-0.03 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P22301 | |
3586 | |
E. coli-derived recombinant human IL-10 Ser19-Asn178 Accession # P22301 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-10 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse IL-10. |
25209 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
IL10 | |
IgG2b | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title