Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$212.08 - $555.88
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | 23738R |
| Dilution | Western Blot 2 ug/mL, Neutralization 5 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB217RSP
![]() |
R&D Systems
MAB217RSP |
25 μg |
Each for $212.08
|
|
|||||
MAB217R100
![]() |
R&D Systems
MAB217R100 |
100 μg |
Each for $555.88
|
|
|||||
Description
IL-10 Monoclonal specifically detects IL-10 in Human samples. It is validated for Western Blot, Neutralization.Specifications
| IL-10 | |
| Western Blot 2 ug/mL, Neutralization 5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| P22301 | |
| 3586 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Detects human IL-10 in direct ELISAs. |
| 23738R | |
| Western Blot, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| IL10 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title