Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $468.50
Specifications
Antigen | IL-10 |
---|---|
Clone | 127107R |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 μg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2172RSP
![]() |
R&D Systems
MAB2172RSP |
25 μg |
Each for $241.00
|
|
|||||
MAB2172R100
![]() |
R&D Systems
MAB2172R100 |
100 μg |
Each for $468.50
|
|
|||||
Description
IL-10 Monoclonal specifically detects IL-10 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
IL-10 | |
ELISA Capture (Matched Antibody Pair) 2-8 μg/mL | |
Monoclonal | |
Supernatant | |
Human | |
P22301 | |
3586 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
127107R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-10, Ser19-Asn178, Accession # P22301 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-10 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title