Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-10 Quantikine HS ELISA Kit
GSA_VA

Catalog No. p-4753925
Click to view available options
Quantity:
1 Plate
6 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Human IL-10 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-10 Quantikine HS ELISA Kit quantifies human IL-10 in serum (200 ul), edta plasma (200 ul), citrate plasma (200 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 0.78 - 50pg/mL
Assay Sensitivity 0.17pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Plasma (EDTA, Citrate), Serum
Specificity Natural and Recombinant Human IL-10
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3586
Quantity 1 Plate
Sample Volume Serum (200uL),EDTA Plasma (200uL),Citrate Plasma (200uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10 mgC126451, interleukin 10, interleukin-10, TGIF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.