Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-10 R alpha Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2742R100UG
This item is not returnable.
View return policy
Description
IL-10R alpha Monoclonal specifically detects IL-10R alpha in Human samples. It is validated for Flow Cytometry.Specifications
IL-10R alpha | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q13651 | |
IL10RA | |
Mouse myeloma cell line NS0-derived and S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-10 R alpha His22-Asn235 Accession # Q13651 | |
100 μg | |
Primary | |
Detects human IL-10 R alpha in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse IL-10 R alpha or recombinant human (rh) IL-10 R beta is observed. In Western blots, no cross-reactivity with rhIL-10 R beta is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
714212 | |
Alexa Fluor 647 | |
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL10R alpha, IL-10R subunit 1, IL-10R1, IL10RA, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3587 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction