Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-11 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
$200.50 - $2020.00
Specifications
Antigen | IL-11 |
---|---|
Clone | 22616 |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
IL-11 Monoclonal specifically detects IL-11 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
IL-11 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P20809 | |
3589 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-11 Pro22-Leu199 Accession # P20809 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-11 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse IL-11 but does not cross-react with recombinant human (rh) IL-6, rhIL-6 R, rhLIF, rhOSM, rhCT-1, rhCNTF, or rhCLC. |
22616 | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin | |
IL11 | |
IgG2b | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title