Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-11 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB218RSP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB218RSP 25 μg
MAB218R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB218RSP Supplier R&D Systems Supplier No. MAB218RSP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-11 Monoclonal antibody specifically detects IL-11 in Human samples. It is validated for Western Blot, Neutralization

Specifications

Antigen IL-11
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 22626R
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P20809
Gene Alias Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-11, Pro22-Leu199, Accession # P20809
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3589
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.