Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-11 R alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $584.50
Specifications
Antigen | IL-11R alpha |
---|---|
Clone | 473143 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
IL-11R alpha Monoclonal specifically detects IL-11R alpha in Human samples. It is validated for Western Blot.Specifications
IL-11R alpha | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q14626 | |
3590 | |
Mouse myeloma cell line NS0-derived recombinant human IL-11 R alpha Cys26-Val363 Accession # Q14626 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-11 R alpha in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with rmIL-11 R alpha or recombinant human IL-6 R alpha is observed. In Western blots, 25% cross-reactivity with recombinant mouse (rm) IL-11 R alpha is observed. |
473143 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AI314697, GP130, IL11R alpha, IL11RA, IL-11RA, Il11ra2, NR1 | |
IL11RA | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title