Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-12/IL-23 p40 MAb (Clone 169516), R&D Systems™
GREENER_CHOICE

Catalog No. MAB6091SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6091SP 25 μg
MAB6091100 100 μg
MAB6091500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB6091SP Supplier R&D Systems Supplier No. MAB6091SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

IL-12/IL-23 p40 Monoclonal specifically detects IL-12/IL-23 p40 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-12/IL-23 p40
Applications Western Blot
Classification Monoclonal
Clone 169516
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P29460
Gene Alias CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40
Gene Symbols IL12B
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-23 p19/p40 heterodimer Ile23-Ser328 Accession # P29460
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3593
Test Specificity Detects human IL-12/IL-23 p40 subunit in direct ELISAs and Western blots. Recognizes IL-12 and IL-23 heterodimers as well as p40 homodimers. Shows 100% cross-reactivity with porcine IL-12/IL-23 p40 and no cross-reactivity with mouse IL-12/IL-23 p40.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.