Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human IL-12 p70 Quantikine HS ELISA Kit
Click to view available options
Quantity:
1 Plate
6 Plates
Description
The Human IL-12 p70 Quantikine HS ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-12 p70/IL-12A in cell culture supernates (200 ul), serum (200 ul), edta plasma (200 ul), heparin plasma (200 ul).
Specifications
Specifications
Assay Range | 0.63 to 40 pg/mL (Cell Culture Supernates), 0.78 to 50 pg/mL (Serum, Heparin Plasma, EDTA Plasma) |
Assay Sensitivity | 0.5pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | Solid Phase Sandwich ELISA |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
Specificity | Natural and Recombinant Human IL-12p70 |
Sufficient For | 1 x 96-well Plate |
Target Species | Human |
For Use With (Application) | ELISA |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction