Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-12 R beta 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB839SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB839SP 25 μg
MAB839 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB839SP Supplier R&D Systems Supplier No. MAB839SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

IL-12 R beta 1 Monoclonal specifically detects IL-12 R beta 1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen IL-12 R beta 1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 69310
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P42701
Gene Alias CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL12R beta 1, IL-12R subunit beta-1, IL12RB, IL12RB1, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454
Gene Symbols IL12RB1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 1 Cys24-Glu540 Accession # P42701
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3594
Test Specificity Detects human IL-12 R beta 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-12 R beta 1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.