Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-12 R beta 1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF839SP
This item is not returnable.
View return policy
Description
IL-12 R beta 1 Polyclonal specifically detects IL-12 R beta 1 in Human samples. It is validated for Western Blot.Specifications
IL-12 R beta 1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL12R beta 1, IL-12R subunit beta-1, IL12RB, IL12RB1, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454 | |
Goat | |
Affinity Purified | |
RUO | |
3594 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
P42701 | |
IL12RB1 | |
Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 1 Cys24-Glu540 Accession # P42701 | |
25 μg | |
Primary | |
Detects human IL-12 R beta 1 in direct ELISAs and Western blots. In these formats, less than 2% cross-reactivity with recombinant mouse IL-12 R beta 1 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction