Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-12 R beta 2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB19591SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB19591SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB19591SP Supplier R&D Systems Supplier No. MAB19591SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-12 R beta 2 Monoclonal specifically detects IL-12 R beta 2 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-12 R beta 2
Applications Western Blot
Classification Monoclonal
Clone 622509
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99665
Gene Alias IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2
Gene Symbols IL12RB2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2 Lys24-Cys622 Accession # Q99665
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3595
Test Specificity Detects human IL-12 R beta 2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human IL-12 R beta 1 or recombinant mouse IL-12 R beta 1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.