Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-13 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2131V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2131V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2131V100 Supplier R&D Systems Supplier No. FAB2131V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-13 Monoclonal specifically detects IL-13 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-13
Applications Flow Cytometry
Classification Monoclonal
Clone 31606
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P35225
Gene Alias ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
Gene Symbols IL13
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-13 Gly35-Asn146 Accession # P35225
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3596
Test Specificity Detects human IL-13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-4 or recombinant mouse IL-13 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.