Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-13 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AF213SP
This item is not returnable.
View return policy
Description
IL-13 Polyclonal specifically detects IL-13 in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
IL-13 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
Goat | |
Affinity Purified | |
RUO | |
3596 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.5-3 ug/mL, Intracellular Staining by Flow Cytometry 5 ug/10^6 cells, CyTOF-ready | |
AAK53823 | |
IL13 | |
E. coli-derived recombinant human IL-13 Gly21-Asn132 Accession # AAK53823 | |
25 μg | |
Primary | |
Detects human IL-13 in direct ELISAs and Western blots. In direct ELISAs, approximately 100% cross-reactivity with recombinant rhesus monkey IL-13 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction