Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-13 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$214.00 - $2150.00
Specifications
Antigen | IL-13 |
---|---|
Clone | 32116 |
Concentration | LYOPH |
Dilution | Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.15-0.75 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Flow Cytometry, Neutralization, CyTOF, ELISA |
Description
IL-13 Monoclonal specifically detects IL-13 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
IL-13 | |
LYOPH | |
Flow Cytometry, Neutralization, CyTOF, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
IL13 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
32116 | |
Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.15-0.75 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P35225 | |
3596 | |
E. coli-derived recombinant human IL-13 Gly35-Asn146 Accession # P35225 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-13 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title