Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-13 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF213
This item is not returnable.
View return policy
Description
IL-13 Polyclonal specifically detects IL-13 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
IL-13 | |
Polyclonal | |
Biotin | |
AAK53823 | |
IL13 | |
E. coli-derived recombinant human IL-13 Gly21-Asn132 Accession # AAK53823 | |
100 μg | |
Primary | |
Detects human IL-13 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant mouse IL-13, recombinant human (rh) IL-13 R alpha 1, and rhIL-13 R alpha 2 is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
Goat | |
Affinity Purified | |
RUO | |
3596 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction