Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-13 R alpha 1 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1462A
This item is not returnable.
View return policy
Description
IL-13R alpha 1 Monoclonal specifically detects IL-13R alpha 1 in Human samples. It is validated for Flow Cytometry.Specifications
IL-13R alpha 1 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL13R alpha 1, IL-13R subunit alpha-1, IL-13Ra, IL13RA, IL13RA1, IL-13RA1, IL-13R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3597 | |
Human | |
Purified |
Flow Cytometry | |
419718 | |
Flow Cytometry 10 uL/10^6 cells | |
P78552 | |
IL13RA1 | |
Mouse myeloma cell line NS0-derived recombinant human IL-13 R alpha 1 Ala27-Thr343 Accession # P78552 | |
100 Tests | |
Primary | |
Detects human IL-13 R alpha 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-13 R alpha 1, recombinant human (rh) IL-13 R alpha 2, rhIL-4 R, rhIL-5 R beta , or rhIL-9 R is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction