Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-13 Protein

Catalog No. 213ILB010
Click to view available options
Quantity:
10 μg
100 μg
25 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-13 Protein is derived from E. coli. The Recombinant Human IL-13 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number AAK53823
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 3596
Molecular Weight (g/mol) 12.3 kDa
Name IL-13
Quantity 10 μg
Source <EM>E. coli</EM>-derived human IL-13 protein Gly21-Asn132
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <0.01 EU / 1 ug of the protein by the LAL method.
Gene Symbol IL13
Conjugate Unconjugated
Cross Reactivity IL-13
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 ug/mL in PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.