Learn More
Gibco™ Human IL-13 variant Recombinant Protein, PeproTech®
Description
PeproTech's genetic variant, termed Recombinant Human IL-13 Variant, is a mature 115 amino acid protein with a substitution of Q for R at position 113. The calculated molecular weight of Recombinant Human IL-13 Variant is 12.6 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P35225 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3596 |
| Molecular Weight (g/mol) | 12.6 kDa |
| Name | Human IL-13 variant |
| Quantity | 100 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.