Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-15 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2471SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2471SP 25 μg
MAB2471100 100 μg
MAB2471500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB2471SP Supplier R&D Systems Supplier No. MAB2471SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 11 publications

IL-15 Monoclonal specifically detects IL-15 in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen IL-15
Applications Western Blot, Neutralization, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 34559
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.1-0.5 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P40933
Gene Alias IL15, IL-15MGC9721, interleukin 15, interleukin-15
Gene Symbols IL15
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-15 Asn49-Ser162 Accession # P40933
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3600
Test Specificity Detects human IL-15 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2, recombinant mouse IL-15, or rhIL-21 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.