Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-15 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 12 publications
$227.50 - $2110.00
Specifications
Antigen | IL-15 |
---|---|
Clone | 34593 |
Dilution | Western Blot 1 ug/mL, Neutralization 0.007-0.042 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Description
IL-15 Monoclonal specifically detects IL-15 in Human samples. It is validated for Western Blot, Neutralization.Specifications
IL-15 | |
Western Blot 1 ug/mL, Neutralization 0.007-0.042 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P40933 | |
3600 | |
E. coli-derived recombinant human IL-15 Asn49-Ser162 Accession # P40933 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-15 in Western blots. In Western blots, no cross-reactivity with recombinant human IL-2 is observed. |
34593 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IL15, IL-15MGC9721, interleukin 15, interleukin-15 | |
IL15 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title