Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-15R alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10900100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10900100 100 μg
MAB10900SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10900100 Supplier R&D Systems Supplier No. MAB10900100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-15R alpha Monoclonal antibody specifically detects IL-15R alpha in Human samples. It is validated for Flow Cytometry

Specifications

Antigen IL-15R alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 2639B
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Accession No. NP_751951
Gene Alias CD215, CD215 antigen, IL15 R alpha, IL-15 R alpha, IL-15 receptor subunit alpha, IL15RA, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human IL-15R alpha, Met1-Lys173, Accession # NP_751951
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3601
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.