Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-16 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC3161U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC3161U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC3161U100 Supplier R&D Systems Supplier No. IC3161U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-16 Monoclonal specifically detects IL-16 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-16
Applications Flow Cytometry
Classification Monoclonal
Clone 70720
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q14005
Gene Alias FLJ16806, FLJ42735, FLJ44234, HsT19289, IL16, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCF, LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16
Gene Symbols IL16
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-16 isoform 1 Met1203-Ser1332 Accession # Q14005
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3603
Test Specificity Detects human IL-16.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.