Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17/IL-17A Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$227.50 - $2070.00
Specifications
Antigen | IL-17/IL-17A |
---|---|
Clone | 41802 |
Dilution | Western Blot 2 ug/mL, Immunohistochemistry 0.5-25 ug/mL, Immunoprecipitation 3 ug/100 ug cell lysate, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3171SP
![]() |
R&D Systems
MAB3171SP |
25 μg |
Each for $227.50
|
|
|||||
MAB3171100
![]() |
R&D Systems
MAB3171100 |
100 μg |
Each for $549.50
|
|
|||||
MAB3171500
![]() |
R&D Systems
MAB3171500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
IL-17/IL-17A | |
Western Blot 2 ug/mL, Immunohistochemistry 0.5-25 ug/mL, Immunoprecipitation 3 ug/100 ug cell lysate, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Mouse | |
Human | |
Q16552 | |
3605 | |
E. coli-derived recombinant human IL-17 Ile20-Ala155 Accession # Q16552 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-17 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) IL-17A/IL-17F heterodimer is observed. No cross-reactivity with recombinant mouse IL-17, recombinant canine Il-17, rhIL-17B, rhIL-17C, rhIL-17D, rhIL-17E, or rhIL-17F is observed. |
41802 | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
IL17A | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title