Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
R&D Systems™ Human IL-17 Quantikine ELISA Kit
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                1 Plate
                            
                        
                            
                                6 Plates
                            
                        
                    Description
The Human IL-17 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-17/IL-17A in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
                
            Specifications
Specifications
| Assay Range | 31.20 - 2000pg/mL | 
| Assay Sensitivity | 15pg/mL | 
| Conjugate | HRP | 
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | 
| Product Type | Solid Phase Sandwich ELISA | 
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | 
| Specificity | Natural and Recombinant Human IL-17 | 
| Sufficient For | 1 x 96-well Plate | 
| Target Species | Human | 
| For Use With (Application) | ELISA | 
| Show More | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction