Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17B Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC1248U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1248U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1248U100 Supplier R&D Systems Supplier No. IC1248U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17B Monoclonal specifically detects IL-17B in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-17B
Applications Flow Cytometry
Classification Monoclonal
Clone 174113
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9UHF5
Gene Alias Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL17B, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
Gene Symbols IL17B
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-17B Gln21-Phe180 Accession # Q9UHF5
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27190
Test Specificity Detects human IL-17B in Western blots and ELISAs. In Western blots, approximately 25% cross-reactivity with recombinant mouse IL-17B is observed and no cross-reactivity with recombinant human (rh) IL-17, rhIL-17C, rhIL-17E or rhIL-17F is observed. In sandwich immunoassays, less than 0.5% cross-reactivity with rmIL-17B, and no cross-reactivity with rhIL-17C, rhIL-17D, rhIL-17E, rhIL-17F, or rhIL-17 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.