Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17B Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $2020.00

Specifications

Antigen IL-17B
Clone 174113
Concentration LYOPH
Dilution Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, Flow Cytometry, CyTOF, ELISA
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1248SP
GREENER_CHOICE
View Documents
R&D Systems
MAB1248SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB1248100
GREENER_CHOICE
View Documents
R&D Systems
MAB1248100
100 μg
Each for $549.50
Only null left
Add to Cart
 
MAB1248500
GREENER_CHOICE
View Documents
R&D Systems
MAB1248500
500 μg
Each for $2,020.00
Only null left
Add to Cart
 
Description

Description

IL-17B Monoclonal specifically detects IL-17B in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

IL-17B
LYOPH
Western Blot, Flow Cytometry, CyTOF, ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL17B, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
IL17B
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
174113
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
Q9UHF5
27190
E. coli-derived recombinant human IL-17B Gln21-Phe180 Accession # Q9UHF5
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human IL-17B in Western blots and ELISAs. In Western blots, approximately 25% cross-reactivity with recombinant mouse IL-17B is observed and no cross-reactivity with recombinant human (rh) IL-17, rhIL-17C, rhIL-17E or rhIL-17F is observed. In sandwich immunoassays, less than 0.5% cross-reactivity with rmIL-17B, and no cross-reactivity with rhIL-17C, rhIL-17D, rhIL-17E, rhIL-17F, or rhIL-17 is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.