Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17C Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1234
This item is not returnable.
View return policy
Description
IL-17C Monoclonal specifically detects IL-17C in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
IL-17C | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q9P0M4.1 | |
IL17C | |
E. coli-derived recombinant human IL-17C His19-Val197 Accession # Q9P0M4.1 | |
500 μg | |
Primary | |
Detects human IL-17C in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-17A, B, D, E or F is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF, ELISA | |
177114 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CX2, Cytokine CX2, IL17C, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27189 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction