Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17D Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC15041U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC15041U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC15041U100 Supplier R&D Systems Supplier No. IC15041U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17D Monoclonal specifically detects IL-17D in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-17D
Applications Flow Cytometry
Classification Monoclonal
Clone 246018
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8TAD2.1
Gene Alias FLJ30846, IL17D, IL-17Dinterleukin 27, IL-22, IL-27interleukin-17D, IL27interleukin-27, interleukin 17D, Interleukin-27
Gene Symbols IL17D
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-17D Ala18-Pro202 Accession # Q8TAD2.1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53342
Test Specificity Detects human IL-17D in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, rhIL-17E, or rhIL-17F is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.