Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17D Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC15041S100UG
This item is not returnable.
View return policy
Description
IL-17D Monoclonal specifically detects IL-17D in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| IL-17D | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8TAD2.1 | |
| IL17D | |
| E. coli-derived recombinant human IL-17D Ala18-Pro202 Accession # Q8TAD2.1 | |
| RUO | |
| 53342 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 246018 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FLJ30846, IL17D, IL-17Dinterleukin 27, IL-22, IL-27interleukin-17D, IL27interleukin-27, interleukin 17D, Interleukin-27 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IL-17D in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, rhIL-17E, or rhIL-17F is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction