Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17D Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB15041SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB15041SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB15041SP Supplier R&D Systems Supplier No. MAB15041SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17D Monoclonal specifically detects IL-17D in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen IL-17D
Applications Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 246018
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8TAD2.1
Gene Alias FLJ30846, IL17D, IL-17Dinterleukin 27, IL-22, IL-27interleukin-17D, IL27interleukin-27, interleukin 17D, Interleukin-27
Gene Symbols IL17D
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-17D Ala18-Pro202 Accession # Q8TAD2.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53342
Test Specificity Detects human IL-17D in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, rhIL-17E, or rhIL-17F is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.