Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17D Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1504SP
This item is not returnable.
View return policy
Description
IL-17D Monoclonal specifically detects IL-17D in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).Specifications
IL-17D | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q8TAD2.1 | |
IL17D | |
E. coli-derived recombinant human IL-17D Ala18-Pro202 Accession # Q8TAD2.1 | |
25 μg | |
Primary | |
Detects human IL-17D in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, rhIL-17E, or rhIL-17F. | |
Human | |
Purified |
Western Blot, Blockade of Receptor-ligand Interaction, ELISA | |
246002 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ30846, IL17D, IL-17Dinterleukin 27, IL-22, IL-27interleukin-17D, IL27interleukin-27, interleukin 17D, Interleukin-27 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
53342 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction