Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17E/IL-25 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1258500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1258500 500 μg
MAB1258SP 25 μg
MAB1258100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1258500 Supplier R&D Systems Supplier No. MAB1258500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for Western Blot, ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen IL-17E/IL-25
Applications Western Blot, ELISA, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 182203
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9H293
Gene Alias IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25
Gene Symbols IL25
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-17E Tyr33-Gly177 Accession # Q9H293
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64806
Test Specificity Detects human IL-17E in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse IL-17E and no cross-reactivity with recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, or rhIL-17F.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.